Learn More
Tocris Bioscience™ Pomalidomide 4'-PEG4-azide
Description
Pomalidomide 4'-PEG4-azide is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG4 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 4-[(14-Azido-3,6,9,12-tetraoxatetradec-1-yl)amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione |
| CAS | 2271036-47-4 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C23H30N6O8 |
| Purity | 0.95 |