Learn More
Tocris Bioscience™ Pomalidomide 4'-PEG1-azide
GREENER_CHOICE
Product Code. 18716076
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. 18716076
Brand: Tocris Bioscience™ 7434/25
Cereblon ligand with PEG linker and terminal azide for onward chemistry
Functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG1 linker with terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-[[2-(2-Azidoethoxy)ethyl]amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione
CAS 2271036-44-1
Quantity 25 mg
Target Degrader Building Blocks
Molecular Formula C17H18N6O5
Purity 0.95