Learn More
SP Bel-Art™ Polygon Magnetic Stirring Bar Spinbar™
Scrapes solids from vessel bottom. High quality PTFE encapsulated magnetic stirring bars ensure efficient mixing with consistent results.
169.00 NOK - 333.00 NOK
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
16418505
|
SP Bel-Art™
F37120-1205 |
12 mm | 5 mm |
171.00 NOK
Pack of 10 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
16438505
|
SP Bel-Art™
F37120-2008 |
20 mm | 8 mm |
221.00 NOK
Pack of 10 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
16468505
|
SP Bel-Art™
F37120-3008 |
30 mm | 8 mm |
276.00 NOK
Pack of 10 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
16478505
|
SP Bel-Art™
F37120-4008 |
40 mm | 8 mm |
333.00 NOK
Pack of 10 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
16448505
|
SP Bel-Art™
F37120-2205 |
22 mm | 5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16458505
|
SP Bel-Art™
F37120-2508 |
25 mm | 8 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16428505
|
SP Bel-Art™
F37120-1506 |
15 mm | 6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16408505
|
SP Bel-Art™
F37120-1006 |
10 mm | 6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Multi-faceted polygon shape with increased surface area creates added turbulence relative to similar sized, smooth cylindrical bars to provide thorough mixing
- Absence of a pivot ring allow the Spinbar to stay close to the bottom of the vessel enabling “scraping” of solids
- Optimal coupling with magnetic spindrive as entire length of stir bar is in close contact
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating; Superior chemical compatibility
- Autoclave safe
Specifications
10/Pk. | |
Polygon |