Learn More
SP Bel-Art™ Polygon Magnetic Stirring Bar Spinbar™
Scrapes solids from vessel bottom. High quality PTFE encapsulated magnetic stirring bars ensure efficient mixing with consistent results.
Brand: SP Bel-Art™ F37120-1006
Additional Details : Weight : 0.45930kg
Description
- Multi-faceted polygon shape with increased surface area creates added turbulence relative to similar sized, smooth cylindrical bars to provide thorough mixing
- Absence of a pivot ring allow the Spinbar to stay close to the bottom of the vessel enabling “scraping” of solids
- Optimal coupling with magnetic spindrive as entire length of stir bar is in close contact
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating; Superior chemical compatibility
- Autoclave safe
Specifications
6 mm | |
10/Pk. | |
10 mm | |
Polygon |