Learn More
SP Bel-Art™ Polygon Magnetic Stirring Bar Spinbar™
Scrapes solids from vessel bottom. High quality PTFE encapsulated magnetic stirring bars ensure efficient mixing with consistent results.
Brand:  SP Bel-Art™ F37120-4008
Additional Details : Weight : 1.96180kg
 View more versions of this product
Product Code. 16478505
  • 333.00 NOK / Pack of 10
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Description
Description
  • Multi-faceted polygon shape with increased surface area creates added turbulence relative to similar sized, smooth cylindrical bars to provide thorough mixing
  • Absence of a pivot ring allow the Spinbar to stay close to the bottom of the vessel enabling “scraping” of solids
  • Optimal coupling with magnetic spindrive as entire length of stir bar is in close contact
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating; Superior chemical compatibility
  • Autoclave safe
Specifications
Specifications
8 mm
10/Pk.
40 mm
Polygon