Learn More
Cole-Parmer™ PolarSafe™ Dewar and Traceable™ Temperature Data Logger Bundles
Shop All Cole-Parmer Products
Click to view available options
Capacity (Metric):
2 L
30 L
35 L
47 L
95 L
115 L
Unit Size:
Each
Description
- Improve sample protection by maintaining specific temperature ranges for extended time periods
- Maintain sample integrity with Traceable™ temperature monitoring
- Data logger is provided with a 2-year NIST-traceable calibration certificate
Specifications
Specifications
| Capacity (Metric) | 47 L |
| Depth (English) | 28.25 in. |
| Depth (Metric) | 71.76 cm |
| Diameter (English) Inner | 5 in. |
| Diameter (English) Outer | 21.3 in. |
| Diameter (Metric) Inner | 12.7 cm |
| Diameter (Metric) Outer | 54.10 cm |
| Material | Aluminum |
Product Content Correction
Product Title
Cole-Parmer™ PolarSafe™ Dewar and Traceable™ Temperature Data Logger Bundles >