Learn More
Grant Instruments™ Platform for PSU-10i Orbital Platform Shaker
Secures tubes to an orbital shaker during shaking applications. Grant Instruments™ Platform for PSU-10i Orbital Shaking Platform holds 88 tubes up to 30mm dia.
Brand:  Grant Instruments™ P16-88
Product Code. 13476458
  • ONLINE EXCLUSIVE PRICE
    4055.00 NOK
    3954.19 NOK / Each
    Save 100.81 NOK
    2% Off
Only null left
Estimated Shipment: 27-01-2026
Description
  • Holds 88 tubes up to 30mm dia.
  • For use with PSU-10i Orbital Platform Shaker
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Up to 88 tubes up to 30mm diameter
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Grant Instruments™ Platform for PSU-10i Orbital Platform Shaker > For Use With: Up to 88 tubes up to 30mm diameter