Learn More
Tocris Bioscience™ Phortress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Phortress is a prodrug of the antitumor agent 5F 203, which acts via binding to aryl hydrocarbon receptors. Induces expression of CYP1A1 and generates adducts in the DNA of sensitive MCF7 and IGROV-1 cells.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | Phortress |
| Purity | >98% |
| Molecular Weight (g/mol) | 459.41 |
| Inhibitors | Phortress |
For Research Use Only