Learn More
Tocris Bioscience™ Phortress
GREENER_CHOICE
Product Code. 18759007
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18759007 10 mg 10mg
18799007 50 mg 50mg
2 options
Product Code. 18759007
Brand: Tocris Bioscience™ 4995/10
Prodrug of the antitumor agent 5F 203
Phortress is a prodrug of the antitumor agent 5F 203, which acts via binding to aryl hydrocarbon receptors. Induces expression of CYP1A1 and generates adducts in the DNA of sensitive MCF7 and IGROV-1 cells.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Product Type Phortress
Purity >98%
Molecular Weight (g/mol) 459.41
Inhibitors Phortress
For Research Use Only