Learn More
Tocris Bioscience™ Phalloidin-FITC
Description
Phalloidin-FITC is a green fluorescent cytoskeleton stain. Binds and labels F-actin. For most fluorescent imaging applications, cells should be fixed and permeablilized. Excitation/emission maxima λ ∽496/516 nm.
We suggest making up a stock solution of this product by dissolving the vial contents in 1.5 mL methanol or DMSO. Please refer to the protocol for further information on using this product.
Specifications
Specifications
| Chemical Name or Material | 7-[(4R)-N5-[[(3',6'-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9'-[9H]xanthen]-5-yl)amino]thioxomethyl]-4-hydroxy-4-methyl-L-ornithine]phalloidin |
| CAS | 915026-99-2 |
| Quantity | 12 μg |
| Target | Fluorescent Actin Probes |
| Molecular Formula | C56H60N10O15S2 |
| Formula Weight | 1177.26 |
| Purity | 90% |