Learn More
Tocris Bioscience™ Pentostatin
GREENER_CHOICE
Product Code. 18782504
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18782504 10 mg 10mg
18722744 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18782504 Supplier Tocris Bioscience™ Supplier No. 2033/10
Adenosine deaminase inhibitor
Irreversible inhibitor of adenosine deaminase (Ki = 2.5 pM). Anticancer agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 53910-25-1
Molecular Formula C11H16N4O4
Specifications
Quantity 10 mg
Formula Weight 268.27
Purity >99%
Molecular Weight (g/mol) 268.27
Chemical Name or Material Pentostatin