Learn More
Manutan Padlock series 65, length: 30 mm, handle Ø: 5 mm,aperture width: 16 mm
Brand:  Manutan A016561
Encompass_Preferred
Padlock series 65, length: 30 mm, handle Ø: 5 mm,aperture width: 16 mm
Product Code. 15645281
  • 155.25 NOK / Each
Only null left
Estimated Shipment: 24-10-2025
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Manutan Padlock series 65, length: 30 mm, handle Ø: 5 mm,aperture width: 16 mm