Learn More
B Braun™ Omnifix Solo, Eccentric Luer-tip, 3-part Syringe Product Code.: 10713047
B Braun™ Omnifix Solo, Eccentric Luer-tip, 3-part Syringe
Product Code. 10713047
3 mL, Pack of 100
Click to view available options
Volume (Metric):
3 mL
10 mL
20 mL
Unit Size:
Pack of 100
Product Code. 10713047
Brand: B Braun™ 4616025V
Use the B Braun™ Omnifix Solo, Eccentric Luer-tip, 3-part Syringe for patient care when for slow aspiration and injection are necessary.
  • Disposable
  • High contrast black graduations
  • Eccentric Leur-tip
Specifications
Certifications/Compliance EN ISO 7886-1, ISO 10993 resp. DIN EN 30993
Material Polypropylene (Plunger), Synthetic Rubber (Stopper)
Color Natural
Description High contrast black graduation
For Use With (Application) For slow aspiration and injection
Latex Free Yes
Product Type Eccentric Luer-tip 3-part Syringe
Disposable Single-use
Graduations 0.1 mL
Quantity 100/Pk.
Volume (Metric) 3 mL
Connection Type Eccentric Luer Tip
Graduated Yes
Sterility Sterile
Product Title
B Braun™ Omnifix Solo, Eccentric Luer-tip, 3-part Syringe >