missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ OGDA

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
OGDA is a green fluorescent D-amino acid. Suitable for labeling peptidoglycans in live bacteria. OGDA is efficiently incorporated into the peptidoglycans of gram-positive and some gram-negative bacteria. Compatible with STED microscopy, OGDA labelled bacteria can be imaged by super-resolution stimulated emission depletion (STED) at a resolution below 100 nm. Photo-stable. Excitation/emission λ ∼501/526 nm.
Specifications
Specifications
| Chemical Name or Material | (R)-2-Amino-3-(2 ',7 '-difluoro-3 ',6 '-dihydroxy-3-oxo-3 H-spiro[isobenzofuran-1,9 '-xanthene]-6-carboxamido)propanoic acid |
| Quantity | 5 mg |
| Target | Fluorescent Probes for Imaging Bacteria |
| Molecular Formula | C24H16F2N2O8 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction