Learn More
Macherey-Nagel™ Nucleodur™ C18 Varioprep VP HPLC Columns Product Code.: 12626009
Only undefined left
Add to basket
Macherey-Nagel™ Nucleodur™ C18 Varioprep VP HPLC Columns
Product Code. 12626009 Shop All Macherey-Nagel Products
250 mm, Each
Click to view available options
Length (Metric):
125 mm
250 mm
Unit Size:
Each
Product Code. 12626009
Brand: Macherey-Nagel™ 762556.100
Only undefined left
Add to basket
Only undefined left
Add to basket
Column Format: Preparative Column; Carbon Load: 18%; Pore Size: 110 Ă…; Column Phase: Reversed Phase
Specifications
Column Format Preparative Column
Diameter (Metric) 10 mm
For Use With Sophisticated Analytical and Preparative Separations of Basic, Neutral and Acidic Pharmaceuticals, Derivatized Amino Acids, Pesticides, Fat-Soluble Vitamins, Aldehydes, Ketones and Phenolic Compounds
Length (Metric) 250 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 ÎĽm
pH 1 to 11
Pore Size 110 Ă…
Surface Area 340 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18 HT
Packing Material Silica
Carbon Load 18%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type Preparative Column
Product Title
Macherey-Nagel™ Nucleodur™ C18 Varioprep VP HPLC Columns >