Learn More
Tocris Bioscience™ NS 3861
Description
NS 3861 is a α3β2 full agonist (EC50 = 1.6 μM). Also α3β4 partial agonist (EC50 = 1 μM). Exhibits minimal activity at α4β2 and α4β4. Displays high affinity for α3β4, α4β4, α3β2 and α4β2 (Ki values are 0.62, 7.8, 25 and 55 nM respectively).
Specifications
Specifications
| Chemical Name or Material | 3-(3-Bromo-2-thienyl)-8-methyl-8-azabicyclo[3.2.1]oct-2-ene fumarate |
| CAS | 216853-60-0 |
| Quantity | 5 mg |
| Target | Additional Nicotinic Receptor Agonists |
| Molecular Formula | C12H14BrNS.C4H4O4 |
| Purity | 0.98 |