Learn More
Non-Return Valve for Vacuum Lines
Designed for use in vacuum lines and fluid handling systems
453.00 NOK - 520.00 NOK
Specifications
Product Type | Non-return Valve |
---|---|
Material | Polypropylene, Neoprene |
For Use With (Application) | For preventing back pressure in filter flasks |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10208114
|
Azlon
NWH902P |
4 to 7 mm |
453.00 NOK
Pack of 5 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
10709155
|
Azlon
NWH906P |
11 to 15 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Neoprene disc valve has no spring and will operate at very low back pressures (NWH905P valve is now manufactured from nitrile rubber)
- Useful for preventing back pressure in filter flasks
- Printed arrow indicates the direction of flow
- Rigid with stepped tapered arms to give firm grip on tubing
- Easily disassembled for cleaning
Specifications
Non-return Valve | |
Polypropylene, Neoprene | |
For preventing back pressure in filter flasks |