Learn More
Non-Return Valve for Vacuum Lines
Designed for use in vacuum lines and fluid handling systems
Brand:  Azlon NWH906P
Additional Details : Weight : 0.28000kg
 View more versions of this product
Product Code. 10709155
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Neoprene disc valve has no spring and will operate at very low back pressures (NWH905P valve is now manufactured from nitrile rubber)
  • Useful for preventing back pressure in filter flasks
  • Printed arrow indicates the direction of flow
  • Rigid with stepped tapered arms to give firm grip on tubing
  • Easily disassembled for cleaning
Specifications
Specifications
Non-return Valve
11 to 15 mm
Polypropylene, Neoprene
For preventing back pressure in filter flasks