Learn More
Non-Return Valve for Vacuum Lines
Designed for use in vacuum lines and fluid handling systems
Brand: Azlon NWH906P
Additional Details : Weight : 0.28000kg
Description
- Neoprene disc valve has no spring and will operate at very low back pressures (NWH905P valve is now manufactured from nitrile rubber)
- Useful for preventing back pressure in filter flasks
- Printed arrow indicates the direction of flow
- Rigid with stepped tapered arms to give firm grip on tubing
- Easily disassembled for cleaning
Specifications
Non-return Valve | |
11 to 15 mm | |
Polypropylene, Neoprene | |
For preventing back pressure in filter flasks |