Learn More
Nitrite Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™ Product Code.: 12962165
  • ONLINE EXCLUSIVE PRICE
    627.00 NOK
    589.47 NOK / 100mL
    Save 37.53 NOK
    5% Off
Nitrite Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
BIOPHARMA
Product Code. p-8014481 Shop All Fisher Chemical Products
100 mL, 100mL
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Product Code. 12962165
Brand: Fisher Chemical A 033 100
  • ONLINE EXCLUSIVE PRICE
    627.00 NOK
    589.47 NOK / 100mL
    Save 37.53 NOK
    5% Off
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Solution Contains H2O
Concentration 1 g/L
Quantity 100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number1134043 Certificate TypeCertificate of Analysis Date23/07/2025 Product CodeA 033 100
Lot Number1127438 Certificate TypeCertificate of Analysis Date03/07/2025 Product CodeA 033 100
Lot Number1125074 Certificate TypeCertificate of Analysis Date19/06/2025 Product CodeA 033 100
Lot Number1065837 Certificate TypeCertificate of Analysis Date04/03/2025 Product CodeA 033 100
Lot Number1026480 Certificate TypeCertificate of Analysis Date04/03/2025 Product CodeA 033 100
Product Content Correction
Product Title
Nitrite Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™ >