missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NF 023
GREENER_CHOICE

Product Code. p-4785221
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18045142

Brand: Tocris Bioscience™ 1240/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective and competitive P2X1 antagonist

A subtype-selective, competitive and reversible P2X1 receptor antagonist. Displays IC50 values of 0.21, 28.9, > 50 and > 100 μM for human P2X1, P2X3, P2X2, and P2X4-mediated responses respectively. Selective over adrenoceptors, histamine and P2Y receptors. Also selectively inhibits the α-subunit of Go/i (EC50 ∽300 nM). Also inhibits DNA-binding activity of HMGA2 (IC50 = 10.63 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104869-31-0
Molecular Formula C35H20N4Na6O21S6

Specifications

Quantity 10 mg
Formula Weight 1162.86
Purity >98%
Molecular Weight (g/mol) 1162.851
Chemical Name or Material NF 023
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.