Learn More
N-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic acid, Millipore™
Description
Buffer substance TES
Specifications
Specifications
| Chemical Name or Material | N-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic acid |
| CAS | 7365-44-8 |
| Synonym | N-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic acid, TES, N-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic acid |
| Quantity | 10 kg |
| Molecular Weight (g/mol) | 229.24 |