missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-PAC Mouse anti-Human, Unconjugated, Clone: 1A5E1, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | N-PAC |
---|---|
Clone | 1A5E1 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889915
|
Proteintech
67340-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16879915
|
Proteintech
67340-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The N-PAC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human N-PAC. This antibody recognizes human, mouse, rat antigen. The N-PAC antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
N-PAC | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
84656 | |
Primary |
1A5E1 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2b |