Learn More
N-PAC Mouse anti-Human, Unconjugated, Clone: 1A5E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67340-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
The N-PAC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human N-PAC. This antibody recognizes human, mouse, rat antigen. The N-PAC antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
N-PAC | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
1A5E1 | |
Unconjugated | |
150 μL | |
84656 | |
IgG2b |