Learn More
N-PAC Mouse anti-Human, Unconjugated, Clone: 1A5E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67340-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16879915
  • 4593.76 NOK / 150µL
Estimated Shipment: 24-05-2024
Description
The N-PAC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human N-PAC. This antibody recognizes human, mouse, rat antigen. The N-PAC antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
N-PAC
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
1A5E1
Unconjugated
150 μL
84656
IgG2b