Learn More
Tocris Bioscience™ N-Acetylcysteine
Product Code. 18747086
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18747086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18747086 Supplier Tocris Bioscience™ Supplier No. 7874/100
  • ONLINE EXCLUSIVE PRICE
    856.00 NOK
    809.55 NOK / 100mg
    Save 46.45 NOK
    5% Off
View all promotions
 Restricted Item
In Stock
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Glutathione (GSH) precursor; maintains stem cell function in culture.
N-Acetylcysteine is a cell-permeable antioxidant and a precursor of reduced glutathione (GSH) (Cat. No. 5219), with anti-inflammatory, mucolytic, and antiviral activities. N-Acetylcysteine can be used in organoid culture media for non-small cell lung and colorectal cancer organoids, expansion media for mouse/human liver and pancreas 3D organoids and LWRN media for colonic 3D organoids from human biopsies. It is a component of S7 medium in the optimized protocol for generation of functional stem cell-derived islets (SC-islets). N-Acetylcysteine can be orally administered.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2R)-2-Acetamido-3-sulfanylpropanoic acid
CAS 616-91-1
Quantity 100 mg
Target Additional Antioxidant Compounds
Molecular Formula C5H9NO3S