Learn More
Multi-element Ionic Chromatography Cation Standard solution, H2O matrix, Fisher chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Description
Multi-element Ionic Chromatography Cation Standard solution containing 4 components; 10mg/kg each of Lithium (Li+), Sodium (Na+), Potassium (K+), Calcium (Ca2+) for Ion Chromatography H2O matrix
Specifications
Specifications
Chemical Name or Material | Four Components Solution |
Packaging | Bottle |
Analyte or Component Names | Lithium, Sodium, Potassium, Calcium |
Physical Form | Liquid |
Concentration | 10mg/Kg of each component |
Type | Multi-component Cationic Standard Solution |
Element | Li, Na, K, Ca |
Solvent or Matrix | Water |
Quantity | 100 mL |
Product Content Correction
Product Title
Multi-element Ionic Chromatography Cation Standard solution, H2O matrix, Fisher chemical™ >