Learn More
Invitrogen™ eBioscience™ Monensin Solution (1000X)
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
- Monensin is an inhibitor of intracellular protein transport
- Incubation of cells in culture with Monensin leads to blockade of protein transport to the Golgi complex (GC) and accumulation of proteins in the endoplasmic reticulum (ER)
- Monensin is effective for enhanced detection of a majority of mouse and human intracellular cytokines; however, it is advised that the investigators evaluate the use and efficacy of this reagent as well as other protein transport inhibitors such as Brefeldin A in their specific assay system
Specifications
Specifications
| Quantity | 1 mL |
| Product Line | eBioscience |
| Physical Form | Liquid |
| Product Type | Inhibitor |