Learn More
Invitrogen™ Pepstatin A, BODIPY™ FL Conjugate
Product Code. 11589196
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11589196 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11589196 Supplier Invitrogen™ Supplier No. P12271
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
For the study of cathepsin D trafficking in cells by fluorescence microscopy
BODIPY™ FL pepstatin A, a green-fluorescent probe, which inhibits cathepsin D in vitro (IC50 approx. 50nM), targets cathepsin D within lysosomes of live cells. Initial studies with this new probe indicate that it is transported to the lysosomes of live cells via an endocytic pathway and that it can be competitively displaced by unlabeled pepstatin A.
Apoptosis, Caspase Activation, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Enzyme and Protein Activity Assays, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 μg
Shipping Condition Room Temperature
Product Line BODIPY
Physical Form Solid
Product Type Inhibitor