Learn More
Corning™ Modified Impeller Assembly Product Code.: 10727993
Estimated Shipment: 16-02-2026
  • ONLINE EXCLUSIVE PRICE
    13530.69 NOK
    13436.63 NOK / Each
    Save 94.06 NOK
    0.70% Off
Only null left
Add to basket
Corning™ Modified Impeller Assembly
Product Code. p-8007929 Shop All Corning Products
Magnetically-Driven 15L Proculture Spinner Flask, Each
Click to view available options
For Use With (Equipment):
Magnetically-Driven 15L Proculture Spinner Flask
Magnetically-Driven 3L Proculture Spinner Flask
Magnetically-Driven 8L Proculture Spinner Flask
Unit Size:
Each
Product Code. 10727993
Brand: Corning™ 401661
  • ONLINE EXCLUSIVE PRICE
    13530.69 NOK
    13436.63 NOK / Each
    Save 94.06 NOK
    0.70% Off
Only null left
Estimated Shipment: 16-02-2026
Add to basket
Only null left
Add to basket
Stainless steel impeller shaft with modified impeller blade for use with probes to create a small bioreactor.
Stainless steel impeller shaft with modified impeller blade for use with probes to create a small bioreactor.
TRUSTED_SUSTAINABILITY
Specifications
Description Impeller shaft with dual bearing
Material Stainless Steel
For Use With (Equipment) Magnetically-Driven 15L Proculture Spinner Flask
Sterility Non-sterile
Product Content Correction
Product Title
Corning™ Modified Impeller Assembly >