Learn More
Corning™ Modified Impeller Assembly
Shop All Corning Products
Click to view available options
For Use With (Equipment):
Magnetically-Driven 15L Proculture Spinner Flask
Magnetically-Driven 3L Proculture Spinner Flask
Magnetically-Driven 8L Proculture Spinner Flask
Unit Size:
Each
Description
Stainless steel impeller shaft with modified impeller blade for use with probes to create a small bioreactor.
Specifications
Specifications
Material | Stainless Steel |
For Use With (Equipment) | Magnetically-Driven 8L Proculture Spinner Flask |
Sterility | Non-sterile |
Product Content Correction
Product Title
Corning™ Modified Impeller Assembly >