missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ML 339

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ML 339 is a potent and selective hCXCR6 antagonist (IC50 = 140 nM); 100-fold less active at the murine CXCR6 receptor (IC50 = 18 μM). Exhibits selectivity over CXCR5, CXCR4, CCR6 and APJ receptors (IC50 >79 μM).
Specifications
Specifications
| Chemical Name or Material | N-[(1 R,3 S,5 S)-9-(2-((2-Chlorophenyl)amino)-2-exoethyl)-9-azabicyclo[3.3.1 ]nonan-3-yl)-3,4,5-trimethoxybenzamide |
| CAS | 2080300-49-6 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 20mM in ethanol |
| Target | Chemokine Receptor Antagonists |
| Molecular Formula | C26H32ClN3O5 |
| Formula Weight | 502 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Show More |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction