Learn More
SP Bel-Art™ Metric Color Micro Spinbar Magnetic Stirring Bars
Micro magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers.
163.00 NOK - 535.00 NOK
Specifications
Autoclavable | Autoclavable |
---|---|
Coating | PTFE, FDA-grade, USP Class VI |
Material | PTFE |
Quantity | 10 Pk. |
Product Code | Brand | Color | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||
16855443
|
SP Bel-Art™
F37123-0603 |
Blue | 6 mm | 3 mm |
232.00 NOK
Pack of 10 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
|||
16855423
|
SP Bel-Art™
F37121-0303 |
Red | 3 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16865423
|
SP Bel-Art™
F37121-0502 |
Red | 5 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16875423
|
SP Bel-Art™
F37121-0603 |
Red | 6 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16885423
|
SP Bel-Art™
F37121-0702 |
Red | 7 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16895423
|
SP Bel-Art™
F37121-0802 |
Red | 8 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16805433
|
SP Bel-Art™
F37121-1003 |
Red | 10 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16815433
|
SP Bel-Art™
F37121-1303 |
Red | 13 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16825433
|
SP Bel-Art™
F37121-1502 |
Red | 15 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16835433
|
SP Bel-Art™
F37122-0202 |
Yellow | 2 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16845433
|
SP Bel-Art™
F37122-0303 |
Yellow | 3 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16855433
|
SP Bel-Art™
F37122-0502 |
Yellow | 5 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16865433
|
SP Bel-Art™
F37122-0603 |
Yellow | 6 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16875433
|
SP Bel-Art™
F37122-0702 |
Yellow | 7 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16885433
|
SP Bel-Art™
F37122-0802 |
Yellow | 8 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16895433
|
SP Bel-Art™
F37122-1003 |
Yellow | 10 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16805443
|
SP Bel-Art™
F37122-1303 |
Yellow | 13 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16815443
|
SP Bel-Art™
F37122-1502 |
Yellow | 15 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16825443
|
SP Bel-Art™
F37123-0202 |
Blue | 2 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16835443
|
SP Bel-Art™
F37123-0303 |
Blue | 3 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16845443
|
SP Bel-Art™
F37123-0502 |
Blue | 5 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16845423
|
SP Bel-Art™
F37121-0202 |
Red | 2 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16865443
|
SP Bel-Art™
F37123-0702 |
Blue | 7 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16875443
|
SP Bel-Art™
F37123-0802 |
Blue | 8 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16885443
|
SP Bel-Art™
F37123-1003 |
Blue | 10 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16895443
|
SP Bel-Art™
F37123-1303 |
Blue | 13 mm | 3 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
16805453
|
SP Bel-Art™
F37123-1502 |
Blue | 15 mm | 2 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
- Available in a variety of colors and sizes
- Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating – Superior Chemical Compatibility
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Specifications
Autoclavable | |
PTFE, FDA-grade, USP Class VI | |
PTFE | |
10 Pk. |