missing translation for 'onlineSavingsMsg'
Learn More

SP Bel-Art™ Metric Color Micro Spinbar Magnetic Stirring Bars

Micro magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers.

Brand:  SP Bel-Art™ F37122-1003

Additional Details : Weight : 0.00570kg

 View more versions of this product

Product Code. 16895433

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Available in a variety of colors and sizes
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Specifications

Specifications

3 mm
Yellow
PTFE
10 mm
PTFE, FDA-grade, USP Class VI
10 Pk.
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers