Learn More
Applied Biosystems™ MagMAX™ Viral/Pathogen Binding Solution Product Code.: 16395025
Applied Biosystems™ MagMAX™ Viral/Pathogen Binding Solution
Product Code. 16395025
550 mL, 1000 reactions
Click to view available options
Quantity:
550 mL
Unit Size:
1000 reactions
Product Code. 16395025
Brand: Applied Biosystems™ A42359
This binding buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.
This binding buffer is included in the MagMAX Viral/Pathogen Nucleic Acid Isolation kits and MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42352, A42356, A42357, and A42358). It is made available separately for applications that require more binding buffer than is provided in the kit.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Sample Type Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Virus, Yeast
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX™
Product Type Viral/Pathogen Binding Solution
Quantity 550 mL
Shipping Condition Room Temperature
Target Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number2837456 Certificate TypeCertificate of Analysis Date29/01/2025 Product CodeA42359
Lot Number2867643 Certificate TypeCertificate of Analysis Date28/01/2025 Product CodeA42359
Lot Number2867836 Certificate TypeCertificate of Analysis Date28/01/2025 Product CodeA42359
Lot Number2898463 Certificate TypeCertificate of Analysis Date28/01/2025 Product CodeA42359
Lot Number2401245 Certificate TypeCertificate of Analysis Date27/01/2025 Product CodeA42359
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Applied Biosystems™ MagMAX™ Viral/Pathogen Binding Solution >