missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LY 2886721 Hydrochloride
GREENER_CHOICE

Product Code. 18748326
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
18748326 50 mg 50mg
18769916 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 18748326

Brand: Tocris Bioscience™ 7081/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective β-secretase (BACE) inhibitor

LY 2886721 Hydrochloride is a potent and selective β-secretase (BACE) inhibitor (IC50 values are 10.2 and 20.3 nM for human BACE2 and BACE1, respectively). Displays >5,000-fold selectivity for BACE over other proteases including cathepsin D, pepsin and renin. Inhibits Aβ1-40 and Aβ1-42 production in cells expressing mutated APP. Reduces hippocampal and cortical Aβ and sAPPβ levels in an Alzheimer's disease mouse model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-[(4 AS,7 AS)-2-Amino-4 A,5-dihydro-4 H-furo[3,4-d][1,3 ]thiazin-7 A(7 H)-yl]-4-fluorophenyl]-5-fluoro-2-pyridinecarboxamide hydrochloride
CAS 1262036-49-6
Quantity 50 mg
Target Additional Synthetic Protease Inhibitors
Molecular Formula C18H16F2N4O2S.HCl
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.