Learn More
Tocris Bioscience™ LY 2119620
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
LY 2119620 is a positive allosteric modulator of M2 and M4 muscarinic receptors. Potentiates activity of agonist at receptor in G protein-coupled functional assay. Cooperativity factor, α = 19.5 and 79.4 for M2 and M4 in presence of ACh, respectively. Binds in extracellular site.
Specifications
Specifications
| Chemical Name or Material | 3-Amino-5-chloro-N-cyclopropyl-4-methyl-6-[2-(4-methyl-1-piperazinyl)-2-oxoethoxy]-thieno[2,3-b]pyridine-2-carboxamide |
| CAS | 886047-22-9 |
| Quantity | 10 mg |
| Target | mAChR M4 Modulators |
| Molecular Formula | C19H24ClN5O3S |
| Formula Weight | 1616 |
| Purity | 98% |