Specifications
| Quantity | 5 g |
| Solubility | Solubility aq. soln.: insoluble. |
| Melting Point | 226.0°C to 233.0°C |
| Formula Weight | 466.96 |
| InChI Key | DMKSVUSAATWOCU-UHFFFAOYNA-N |
| SMILES | CCOC(=O)OC1(CCC2C3CCC4=CC(=O)C=CC4(C)C3C(O)CC12C)C(=O)OCCl |
| IUPAC Name | chloromethyl 1-[(ethoxycarbonyl)oxy]-10-hydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthrene-1-carboxylate |
| Molecular Weight (g/mol) | 466.96 |
| Chemical Name or Material | Loteprednol etabonate |