Learn More
Fisherbrand™ Lab Thermometer
Product Code. 11587273 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +110°C
-10°C to +150°C
-10°C to +50°C
-20°C to +110°C
-20°C to +150°C
Subdivision:
0.5°C
1°C
Immersion Type:
Partial Immersion
Total Immersion
Length (Metric):
155 mm
305 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Subdivision Immersion Type Length (Metric) Unit Size
11587273 -20°C to +110°C 1°C Total Immersion 305 mm Each
11597273 -20°C to +150°C 1°C Total Immersion 305 mm Each
31488169 -10°C to +50°C 1°C Total Immersion 155 mm -
11517283 -10°C to +110°C 1°C Total Immersion 155 mm Each
11551993 -10°C to +50°C 0.5°C Partial Immersion 305 mm Each
11561993 -10°C to +110°C 1°C Partial Immersion 305 mm Each
11571993 -10°C to +150°C 1°C Partial Immersion 305 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11587273
In Stock
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen
A general purpose/stirring thermometer for a wide application of laboratory needs.
  • With amber graduations
  • Non-roll fitting
  • Reinforced bulb for stirring
Spezifikation
For Use With (Application) General use
Immersion Type Total Immersion
Length (Metric) 305 mm
Thermometer Fill Red Spirit
Product Type Liquid Filled Thermometer
Max. Temperature (Metric) 110°C
Min. Temperature (Metric) -20°C
Temperature Range (Metric) -20°C to +110°C
Subdivision 1°C