Learn More
Buerkle™ LDPE Lid for Rectangular Container
Product Code. p-8654006 Shop All Bürkle Products
Click to view available options
For Use With (Equipment):
48 L Container
50 L Container
Unit Size:
Each
Product Code. 10125903
Brand: Buerkle™ 34400481
Buerkle™ LDPE Lid for Rectangular Container is suitable for use with Buerkle™ Rectangular Containers.
  • Linear LDPE
  • Translucent
  • For use with rupture proof rectangular containers
TRUSTED_SUSTAINABILITY
Specifications
Product Type Container Lid
Material LDPE
Shape Rectangular
Closure Type Lid
Color Transparent
For Use With (Equipment) 48 L Container