Learn More
Labconco™ Fast-Freeze™ Flask Adapter
Fast-Freeze™ Flask Adapter for connecting Fast-Freeze™ flasks to freeze-dryer valves.
Brand:  Labconco™ 7546000
583.16 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10491425
  • 600.00 NOK / Each
Only null left
Estimated Shipment: 22-10-2025
Description
For connecting Fast-Freeze™ flasks to freeze-dryer valves
  • 40 and 80 mL flasks take size 0.5 in. or 0.5 to 0.75 in.
  • Larger flasks fit size 0.75 in. or large end of 0.5 to 0.75 in.
Specifications
Specifications
Flask Adapter
Borosilicate Glass
For connecting fast freeze™ flasks to freeze-dryer valves
0.5 to 0.75 in.
0.75 in.
3.25 in.
0.5 lb.
40, 80 mL
45° bend
Fast freeze™ flask
1.27 to 1.9 cm
1.905 cm
8.26 cm
0.2 kg
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Labconco™ Fast-Freeze™ Flask Adapter > Material: Glass, Style: 45° Bend, Diameter: 0.5 to 0.75 in.