missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Asparagine, 99% [Anhydrous], MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 132.1 |
| Grade | Cell Culture |
| Physical Form | Crystals |
| Chemical Name or Material | L-Asparagine, 99% [Anhydrous] |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Asparagine, 99% [Anhydrous], MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction
