missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies KU0060648

Product Code. p-7217642
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
This item is not returnable. View return policy

Product Code. 17110322

Brand: STEMCELL Technologies 74062

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NHEJ pathway inhibitor; Inhibits DNA-dependent protein kinase (DNA-PK)

  • KU0060648 is an ATP-competitive and highly potent inhibitor of DNA-dependent protein kinase (DNA-PK), with IC50 values of 19 nM in MCF7 human breast cancer cells and 170 nM in SW620 human colon cancer cells
  • KU0060648 also inhibits phosphoinositide 3-kinase (PI3K) with IC50 values of 39 nM in MCF7 cells and > 10 μM in SW620 cells, which suggests a cell-dependent mechanism (Munck et al.)
  • Reduces the frequency of non-homologous end joining (NHEJ) and increases the efficiency of homology-directed repair (HDR) in CRISPR-Cas9 genome editing (Robert et al.)
  • Inhibits tumor growth and sensitizes human cancer cells to DNA double-strand-break (DSB)-inducing therapies (chemo- or radio-therapy) by inhibiting both DNA-PK and PI3K and preventing the execution of DSB repair (Chen et al.; Dietlein et al.; Munck et al.)

Specifications

CAS 881375-00-4
Quantity 10 mg
Molecular Formula C33H34N4O4S
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.