Learn More
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures Product Code.: 11863003
Estimated Shipment: 23-03-2026
Only null left
Add to basket
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures
Product Code. 11863003
0.5 dr., 8-425, Pack of 200
Click to view available options
Volume (English):
0.5 dr.
1 dr.
1.5 dr.
10 dr.
2 dr.
5 dr.
Closure Size:
13-425
15-425
24-400
8-425
Unit Size:
Pack of 200
Product Code. 11863003
Brand: DWK Life Sciences 609121235
Only null left
Estimated Shipment: 23-03-2026
Add to basket
Only null left
Add to basket
Manufactured from amber 33 expansion borosilicate glass
  • Amber vials offer protection against harmful light and UV rays for light sensitive products.
TRUSTED_SUSTAINABILITY
Specifications
Color Amber
Diameter (Metric) Outer 12 mm
Material Borosilicate Glass
Volume (Metric) 1.8 mL
Thread Style Screw
Volume (English) 0.5 dr.
Closure Type Screw Cap
Height (Metric) 35 mm
Certifications/Compliance USP Type I
Closure Size 8-425
Product Content Correction
Product Title
DWK Life Sciences Kimble™ AmberScrew Thread Vials without Closures >