Learn More
DWK Life Sciences KECK™ Flat sealing
Spare part for KECK™ Adapter KA.
535.00 NOK - 1940.00 NOK
Product Code | Brand | Diameter (Metric) Outside | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outside | Material | Price | Quantity & Availability | ||||
11706319
|
DWK Life Sciences
286341506 |
12 mm | EPDM |
1065.00 NOK
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
15824698
|
DWK Life Sciences
286341009 |
16 mm | Viton |
1940.00 NOK
Pack of 100 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
15371971
|
DWK Life Sciences
286341609 |
16 mm | EPDM |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The flat gasket as spare part for KECK™ Adapter KA is available in the materials VITON and EPDM.
- The flat gasket for KECK™ Adapter KA is available in the materials VITON and EPDM.
Specifications
Sealing | |
100/Pk. |