Learn More
DWK Life Sciences KECK™ Flat sealing
Spare part for KECK™ Adapter KA.
Brand: DWK Life Sciences 286341009
1843.00 NOK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 80.00000kg
Description
The flat gasket as spare part for KECK™ Adapter KA is available in the materials VITON and EPDM.
- The flat gasket for KECK™ Adapter KA is available in the materials VITON and EPDM.
Specifications
8.5 mm | |
100/Pk. | |
16 mm |
Sealing | |
Viton |