Learn More
Tocris Bioscience™ K34c
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
K34c is a potent and selective α5β1 integrin inhibitor (IC50 = 3.1 nM). K34c inhibits cell survival and migration, inhibits p53-dependent senescence induced by Temozolomide (Cat. No. 2706) or Ellipticine (Cat. No. 3357) and promotes apoptosis in U87MG cells. K34c also prevents TGFβ-induced infiltration in vivo and reduces cell adhesion on fibronectin.
Specifications
Specifications
| Chemical Name or Material | N-(2,6-Dimethylbenzoyl)-O-[3-(2-pyridinylamino)propyl]-L-tyrosine |
| CAS | 939769-93-4 |
| Quantity | 5 mg |
| Target | Integrin Receptor Inhibitors |
| Molecular Formula | C26H29N3O4 |
| Purity | 0.98 |