missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JNJ 10198409
GREENER_CHOICE

Product Code. p-7199111
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18776356

Brand: Tocris Bioscience™ 6976/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent PDGFRα and PDGFRβ inhbitor; also inhibits c-Abl, Lck, c-Src and Fyn kinases

JNJ 10198409 is a potent platelet-derived growth factor receptor (PDGFR) inhibitor (IC50 values are 4.2 and 45 nM for PDGFRβ and PDGFRα, respectively). Also inhibits c-Abl, Lck, c-Src and Fyn kinases (IC50 values are 22, 100, 185 and 378 nM respectively). Displays antiproliferative activity in multiple tumor cell lines. Accelerates down regulation of fibroblast genes in reprogramming of human fibroblasts into functional cardiomyocytes, within 9C cocktail.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(3-Fluorophenyl)-1,4-dihydro-6,7-dimethoxyindeno[1,2-c]pyrazol-3-amine
CAS 627518-40-5
Quantity 10 mg
Target PDGF Receptor Inhibitors
Molecular Formula C18H16FN3O2
Formula Weight 2273
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.