missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies IWP-4
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- Inhibitor of WNT Production-4 (IWP-4) inhibits WNT signaling by inactivating Porcupine, a membrane-bound O-acyltransferase responsible for palmitoylating WNT proteins, which is essential for WNT signaling ability and secretion (Chen et al.) IWP-4 impairs WNT pathway activity in vitro with an IC50 value of 25 nM (Chen et al.)
- Promotes cardiomyocyte differentiation in human pluripotent stem cells after treatment with CHIR99021 (Lian et al.), 2012, 2013; Sequiera et al.)
- Promotes cardiomyocyte differentiation in human embryonic stem cells following primitive streak induction with BMP4 and Activin A (Hudson et al.)
Specifications
Specifications
| CAS | 686772-17-8 |
| Synonym | Inhibitor of Wnt Production-4 |
| Quantity | 10 mg |
| Molecular Formula | C23H20N4O3S3 |
| Purity | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction