Learn More
Tocris Bioscience™ IOX 2
GREENER_CHOICE
Product Code. 18064972
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
Product Code. 18064972
Brand: Tocris Bioscience™ 4451/50
Potent, selective HIF-1α prolyl hydroxylase-2 (PHD2) inhibitor
Potent inhibitor of HIF-1α prolyl hydroxylase-2 (PHD2) (IC50 = 21 nM). Displays over 100-fold selectivity for PHD2 over factor inhibiting HIF-1 (FIH-1) and histone demethylases JMJD2A, JMJD2C, JMJD2E and JMJD3. Cell permeable.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 931398-72-0
Molecular Formula C19H16N2O5
Specifications
Quantity 50 mg
Formula Weight 352.34
Purity >99%
Molecular Weight (g/mol) 352.346
Chemical Name or Material IOX 2