missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Integrin alpha 5 beta 1 Antibody (BMC5), Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
2170.00 NOK - 6650.00 NOK
Specifications
| Antigen | Integrin alpha 5 beta 1 |
|---|---|
| Clone | BMC5 |
| Dilution | Western Blot, Flow Cytometry 10ul/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2 uL, Immunohistochemistry-Frozen 1:10-1:500, Functional 10ul/ml |
| Applications | Functional Assay, Immunohistochemistry (Frozen), Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18146823
|
Novus Biologicals
NBP2-29788-0.025ML |
0.025 mL |
2170.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18767353
|
Novus Biologicals
|
6650.00 NOK
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Integrin alpha 5 beta 1 Monoclonal specifically detects Integrin alpha 5 beta 1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, Immunohistochemistry-Paraffin (Negative).Specifications
| Integrin alpha 5 beta 1 | |
| Western Blot, Flow Cytometry 10ul/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2 uL, Immunohistochemistry-Frozen 1:10-1:500, Functional 10ul/ml | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with No Preservative | |
| 3678 | |
| Mouse embryonal carcinoma P19 cells with myeloma P3X63Ag9.653 cells. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| BMC5 | |
| Functional Assay, Immunohistochemistry (Frozen), Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P08648 | |
| ITGA5 | |
| IgG2b κ | |
| Unpurified | |
| Antibody was raised against mouse integrin VLA-5 (alpha5beta1), a fibronectin receptor. The antibody was prepared by fusion of spleen cells from a Fisher rat immunized with mouse embryonal carcinoma P19 cells with myeloma P3X63Ag9.653 cells. Cell lines tested to express alpha5beta1, include: P19 cells, mouse melanoma B16F1 cells, mouse squamous carcoma SCC cells and thymocytes. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title